COM964466222 Combi-Blocks, Inc.

1-(2-Methanesulfonyloxyethyl)-2-chlorobenzene

  • MDL number MFCD25963245
  • Cas Number : 40759-45-3
  • Molecular Weight : 234.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8903

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8903
Availability details for
Quantity
  • Availability
  • Add to Cart