COM964466304 Combi-Blocks, Inc.

4-(Cyclohex-1-en-1-yl)-3-phenyl-1h-pyrazol-5-amine

  • MDL number MFCD30531309
  • Cas Number : 2201057-79-4
  • Molecular Weight : 239.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9140

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9140
Availability details for
Quantity
  • Availability
  • Add to Cart