COM964466309 Combi-Blocks, Inc.

3-(Pyrimidin-2-yl)-1,2,4-thiadiazol-5-amine

  • MDL number MFCD12093748
  • Cas Number : 138588-24-6
  • Molecular Weight : 179.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9156

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9156
Availability details for
Quantity
  • Availability
  • Add to Cart