COM964466376 Combi-Blocks, Inc.

2-Fluoro-6-nitropyridine

  • MDL number MFCD11845554
  • Cas Number : 1082042-27-0
  • Molecular Weight : 142.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9362
Availability details for
Quantity
  • Availability
  • Add to Cart