COM964466389 Combi-Blocks, Inc.

1H-Indene-1-carboxylic acid

  • MDL number MFCD00093739
  • Cas Number : 1544842-47-8
  • Molecular Weight : 160.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9391

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9391
Availability details for
Quantity
  • Availability
  • Add to Cart