COM964466415 Combi-Blocks, Inc.

4-(2-(Benzyloxy)ethoxy)-3-methoxybenzoic acid

  • MDL number MFCD29917092
  • Cas Number : 1881167-15-2
  • Molecular Weight : 302.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9465

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9465
Availability details for
Quantity
  • Availability
  • Add to Cart