COM964466430 Combi-Blocks, Inc.

(R)-N-(1-(4-Bromothiophen-2-yl)ethylidene)-2-methylpropane-2-sulfinamide

  • MDL number MFCD30472126
  • Cas Number : 1310877-37-2
  • Molecular Weight : 308.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9503

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9503
Availability details for
Quantity
  • Availability
  • Add to Cart