COM964466431 Combi-Blocks, Inc.

1-Benzyl-3-tert-butyl-1h-pyrazol-5-amine

  • MDL number MFCD08442469
  • Cas Number : 866255-96-1
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9505

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9505
Availability details for
Quantity
  • Availability
  • Add to Cart