COM964466492 Combi-Blocks, Inc.

3-(4-Bromo-3-fluorophenoxy)propane-1,2-diol

  • MDL number MFCD24315996
  • Cas Number : 1548539-95-2
  • Molecular Weight : 265.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9674

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9674
Availability details for
Quantity
  • Availability
  • Add to Cart