COM964466510 Combi-Blocks, Inc.

Dimethyl 2,2'-((2-((tert-butoxycarbonyl)amino)ethyl)azanediyl)diacetate

  • MDL number MFCD29917059
  • Cas Number : 212143-39-0
  • Molecular Weight : 304.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9729

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9729
Availability details for
Quantity
  • Availability
  • Add to Cart