COM964466560 Combi-Blocks, Inc.

2,6-Dibromo-3-fluorobenzoic acid

  • MDL number MFCD28784652
  • Cas Number : 1806327-84-3
  • Molecular Weight : 297.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9873

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9873
Availability details for
Quantity
  • Availability
  • Add to Cart