COM964466639 Combi-Blocks, Inc.

Methyl 1-ethylpyrrolidine-2-carboxylate

  • MDL number MFCD07776953
  • Cas Number : 107599-40-6
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6509

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6509
Availability details for
Quantity
  • Availability
  • Add to Cart