COM964466649 Combi-Blocks, Inc.

2,7-Dibromo-9H-fluoren-4-ol

  • MDL number MFCD30834474
  • Cas Number : 2140305-35-5
  • Molecular Weight : 340.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6558

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6558
Availability details for
Quantity
  • Availability
  • Add to Cart