COM964466655 Combi-Blocks, Inc.

4-(1-Methyl-4-hydroxypiperidin-4-yl)phenylboronic acid pinacol ester, HCl

  • MDL number MFCD30834477
  • Cas Number : 2377611-23-7
  • Molecular Weight : 353.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6589

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6589
Availability details for
Quantity
  • Availability
  • Add to Cart