COM964466664 Combi-Blocks, Inc.

1-(tert-Butoxycarbonyl)-1h-pyrrole-2-carboxylic acid

  • MDL number MFCD02094480
  • Cas Number : 117657-40-6
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5046

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5046
Availability details for
Quantity
  • Availability
  • Add to Cart