COMH039FD35E Combi-Blocks, Inc.

N-(2-Aminoethyl)-2-fluorobenzamide

  • MDL number MFCD04356335
  • Cas Number : 152535-12-1
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2531

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2531
Availability details for
Quantity
  • Availability
  • Add to Cart