COMH039FD362 Combi-Blocks, Inc.

1-(2-Fluoro-3-methoxy-6-nitrophenyl)ethanone

  • MDL number MFCD09833146
  • Cas Number : 948553-47-7
  • Molecular Weight : 213.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-fluoro-3-methoxy-6-nitrophenyl)ethan-1-one
Inchi Code 1S/C9H8FNO4/c1-5(12)8-6(11(13)14)3-4-7(15-2)9(8)10/h3-4H,1-2H3
Inchi Key MYAJTCUTOOXOPG-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6260

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6260
Availability details for
Quantity
  • Availability
  • Add to Cart