COMH039FD363 Combi-Blocks, Inc.

5-Bromo-2-fluoro-3-methylaniline hydrochloride

  • MDL number MFCD31543945
  • Cas Number : 2140326-17-4
  • Molecular Weight : 240.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6261

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6261
Availability details for
Quantity
  • Availability
  • Add to Cart