COMH039FD365 Combi-Blocks, Inc.

N-[(5-Fluoro-2-nitrophenyl)methyl]acetamide

  • MDL number MFCD31543946
  • Cas Number : 2140316-94-3
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6263

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6263
Availability details for
Quantity
  • Availability
  • Add to Cart