COMH039FD433 Combi-Blocks, Inc.

7-Bromo-3-methylquinoxalin-2(1H)-one

  • MDL number MFCD14859917
  • Cas Number : 103095-19-8
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-2189

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-2189
Availability details for
Quantity
  • Availability
  • Add to Cart