COMH039FD476 Combi-Blocks, Inc.

[3-(4-Methoxyphenyl)propyl](methyl)amine

  • MDL number MFCD07374124
  • Cas Number : 83986-67-8
  • Molecular Weight : 179.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6630

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6630
Availability details for
Quantity
  • Availability
  • Add to Cart