COMH042332B8 Combi-Blocks, Inc.

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]isoxazole

  • MDL number MFCD09027764
  • Cas Number : 837392-66-2
  • Molecular Weight : 245.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2766
Availability details for
Quantity
  • Availability
  • Add to Cart