COMH0423341A Combi-Blocks, Inc.

3,4-Dihydro-2h-1,5-benzodioxepine-7-carboxylic acid

  • MDL number MFCD00276310
  • Cas Number : 20825-89-2
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3871

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3871
Availability details for
Quantity
  • Availability
  • Add to Cart