COMH04233427 Combi-Blocks, Inc.

Zinc fluoride

  • MDL number MFCD00011298
  • Cas Number : 7783-49-5
  • Molecular Weight : 103.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4286

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4286
Availability details for
Quantity
  • Availability
  • Add to Cart