COMH0423363A Combi-Blocks, Inc.

(2-Phenyl-1,3-thiazol-5-yl)methanol

  • MDL number MFCD09694149
  • Cas Number : 859485-91-9
  • Molecular Weight : 191.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4903

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4903
Availability details for
Quantity
  • Availability
  • Add to Cart