COMH04233CA2 Combi-Blocks, Inc.

Ginkgolide c

  • MDL number MFCD06642603
  • Cas Number : 15291-76-6
  • Molecular Weight : 440.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6473

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6473
Availability details for
Quantity
  • Availability
  • Add to Cart