COMH04233CAE Combi-Blocks, Inc.

1-(2-Chlorothiazol-5-yl)ethanone

  • MDL number MFCD11847548
  • Cas Number : 885229-41-4
  • Molecular Weight : 161.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6544

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6544
Availability details for
Quantity
  • Availability
  • Add to Cart