COMH04233D0E Combi-Blocks, Inc.

Methyl 5-amino-1h-benzo[d]imidazole-2-carboxylate hydrochloride

  • MDL number MFCD30530464
  • Cas Number : 2044704-79-0
  • Molecular Weight : 227.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7301

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7301
Availability details for
Quantity
  • Availability
  • Add to Cart