COMH04233D40 Combi-Blocks, Inc.

2-({3-[(2-hydroxyethyl)sulfanyl]propyl}sulfanyl)ethanol

  • MDL number MFCD01098524
  • Cas Number : 16260-48-3
  • Molecular Weight : 196.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7672

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7672
Availability details for
Quantity
  • Availability
  • Add to Cart