COMH04233D4E Combi-Blocks, Inc.

4-methyl-N-(4-nitrophenyl)benzenesulfonamide

  • MDL number MFCD00129916
  • Cas Number : 734-25-8
  • Molecular Weight : 292.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7784

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7784
Availability details for
Quantity
  • Availability
  • Add to Cart