COMH04233DAE Combi-Blocks, Inc.

N-dodecylaniline

  • MDL number MFCD00026570
  • Cas Number : 3007-74-7
  • Molecular Weight : 261.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8458

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8458
Availability details for
Quantity
  • Availability
  • Add to Cart