COMH04233EE3 Combi-Blocks, Inc.

2-(Bis-ethylsulfanyl-methylene)-malononitrile

  • MDL number MFCD01756906
  • Cas Number : 18771-38-5
  • Molecular Weight : 198.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QW-0172

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QW-0172
Availability details for
Quantity
  • Availability
  • Add to Cart