COMH04234005 Combi-Blocks, Inc.

2,6-Dichloro-n,n-dimethylpyrimidin-4-amine

  • MDL number MFCD09746252
  • Cas Number : 117077-93-7
  • Molecular Weight : 192.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QW-0896

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QW-0896
Availability details for
Quantity
  • Availability
  • Add to Cart