COMH042340C6 Combi-Blocks, Inc.

2-(3-Bromobicyclo[1.1.1]pentan-1-yl)-1-phenylethan-1-one

  • MDL number MFCD30719361
  • Cas Number : 2102408-90-0
  • Molecular Weight : 265.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QW-1373

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QW-1373
Availability details for
Quantity
  • Availability
  • Add to Cart