COMH04234474 Combi-Blocks, Inc.

2-tert-Butyl 8a-methyl (8ar)-6-oxo-1,2,3,4,6,7,8,8a-octahydroisoquinoline-2,8a-dicarboxylate

  • MDL number MFCD29922376
  • Cas Number : 445312-74-3
  • Molecular Weight : 309.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QW-3624

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QW-3624
Availability details for
Quantity
  • Availability
  • Add to Cart