COMH042344DC Combi-Blocks, Inc.

4,7-Dihydro-3h-imidazo[4,5-d][1,3]diazepin-8-one

  • MDL number MFCD28506195
  • Cas Number : 72079-77-7
  • Molecular Weight : 150.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QW-3855

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QW-3855
Availability details for
Quantity
  • Availability
  • Add to Cart