COMH04234579 Combi-Blocks, Inc.

Dibutyldimethoxytin

  • MDL number MFCD00008345
  • Cas Number : 1067-55-6
  • Molecular Weight : 295.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QW-4237

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QW-4237
Availability details for
Quantity
  • Availability
  • Add to Cart