COMH0423494C Combi-Blocks, Inc.

4-(2-Chloroethyl)benzamide

  • MDL number MFCD20637141
  • Cas Number : 181115-67-3
  • Molecular Weight : 183.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QW-6701

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QW-6701
Availability details for
Quantity
  • Availability
  • Add to Cart