COMH042349CB Combi-Blocks, Inc.

N-Benzyl-2-chloroquinazolin-4-amine

  • MDL number MFCD03691634
  • Cas Number : 157864-32-9
  • Molecular Weight : 269.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QW-7005

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QW-7005
Availability details for
Quantity
  • Availability
  • Add to Cart