COMH04234A56 Combi-Blocks, Inc.

T-Boc-n-amido-peg5-ms

  • MDL number MFCD31536757
  • Cas Number : 2128735-28-2
  • Molecular Weight : 415.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QW-7331

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QW-7331
Availability details for
Quantity
  • Availability
  • Add to Cart