COMH04234A94 Combi-Blocks, Inc.

(3R,4R)-3-Methylpiperidin-4-ol hydrochloride

  • MDL number MFCD29060204
  • Cas Number : 955028-91-8
  • Molecular Weight : 151.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QW-7473

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QW-7473
Availability details for
Quantity
  • Availability
  • Add to Cart