COMH04234ADC Combi-Blocks, Inc.

1-(Trans-4-((tert-butyldimethylsilyl)oxy)cyclohexyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD22572246
  • Cas Number : 1257997-17-3
  • Molecular Weight : 406.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QW-7632

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QW-7632
Availability details for
Quantity
  • Availability
  • Add to Cart