COMH04234D82 Combi-Blocks, Inc.

6-Bromo-n-methylpyrid-2-ylmethylamine

  • MDL number MFCD08690325
  • Cas Number : 675109-37-2
  • Molecular Weight : 201.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QW-9237

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QW-9237
Availability details for
Quantity
  • Availability
  • Add to Cart