COMH04234FA1 Combi-Blocks, Inc.

1-[(2,4-Dichlorobenzyl)sulfonyl]piperidine-4-carboxylic acid

  • MDL number MFCD28954580
  • Cas Number : 1858255-09-0
  • Molecular Weight : 352.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0242

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0242
Availability details for
Quantity
  • Availability
  • Add to Cart