COMH04234FA9 Combi-Blocks, Inc.

(1-([3-(3-Methylphenyl)-1-phenyl-1h-pyrazol-4-yl]carbonyl)piperidin-4-yl)acetic acid

  • MDL number MFCD12028308
  • Cas Number : 1172417-82-1
  • Molecular Weight : 403.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0250

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0250
Availability details for
Quantity
  • Availability
  • Add to Cart