COMH04234FF5 Combi-Blocks, Inc.

tert-Butyl 4-(benzoylamino)-3,4-dihydroisoquinoline-2(1h)-carboxylate

  • MDL number MFCD26792692
  • Cas Number : 1785764-68-2
  • Molecular Weight : 352.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0326
Availability details for
Quantity
  • Availability
  • Add to Cart