COMH0423503C Combi-Blocks, Inc.

2-Chloro-1-(1,2,5-trimethyl-1h-indol-3-yl)ethanone

  • MDL number MFCD12027482
  • Cas Number : 1134334-65-8
  • Molecular Weight : 235.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0397

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0397
Availability details for
Quantity
  • Availability
  • Add to Cart