COMH0423508E Combi-Blocks, Inc.

2-Bromo-n-(2-phenylethyl)butanamide

  • MDL number MFCD01615489
  • Cas Number : 1119453-00-7
  • Molecular Weight : 270.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0479

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0479
Availability details for
Quantity
  • Availability
  • Add to Cart