COMH04235092 Combi-Blocks, Inc.

5-(Piperidin-1-ylmethyl)-2-furoic acid hydrochloride

  • MDL number MFCD00573792
  • Cas Number : 301353-36-6
  • Molecular Weight : 245.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0483
Availability details for
Quantity
  • Availability
  • Add to Cart