COMH042350E5 Combi-Blocks, Inc.

Methyl n-(2-methylbenzoyl)glycinate

  • MDL number MFCD01213455
  • Cas Number : 1207-34-7
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0566

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0566
Availability details for
Quantity
  • Availability
  • Add to Cart